Anti-OGT Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41604-100EA 0 EUR
BIRBORB41604-100
Anti-OGT Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: OGT (O-linked N-acetylglucosamine (GlcNAc) transferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR