Anti-DYNC1I2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB41747-100
BIRBORB41747-100EA 0 EUR
BIRBORB41747-100
Anti-DYNC1I2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: DYNC1I2 (dynein, cytoplasmic 1, intermediate chain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR