Anti-ERCC1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Biorbyt
ORB41788-100
BIRBORB41788-100EA 0 EUR
BIRBORB41788-100
Anti-ERCC1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ERCC1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR