Anti-PGAM1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41872-100EA 0 EUR
BIRBORB41872-100
Anti-PGAM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PGAM1 (phosphoglycerate mutase 1 (brain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR