Anti-ALKBH1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB41896-100
BIRBORB41896-100EA 0 EUR
BIRBORB41896-100
Anti-ALKBH1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: ALKBH1 (alkB, alkylation repair homolog 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR