Anti-HSPD1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43289-100EA 0 EUR
BIRBORB43289-100
Anti-HSPD1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR