Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB43309-100EA 0 EUR
BIRBORB43309-100
Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR