Anti-ZW10 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB43316-100
BIRBORB43316-100EA 0 EUR
BIRBORB43316-100
Anti-ZW10 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 1:500

Type: Primary
Antigen: ZW10 (zw10 kinetochore protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR