Anti-DYKDDDDK Tag Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43330-100EA 0 EUR
BIRBORB43330-100
Anti-DYKDDDDK Tag Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR