Anti-EIF2B1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB43346-100
BIRBORB43346-100EA 0 EUR
BIRBORB43346-100
Anti-EIF2B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EIF2B1 (eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR