Anti-IL10RA Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB43352-100
BIRBORB43352-100EA 0 EUR
BIRBORB43352-100
Anti-IL10RA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR