Anti-CTNNB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43357-100EA 0 EUR
BIRBORB43357-100
Anti-CTNNB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR