Anti-CD28 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB43726-100
BIRBORB43726-100EA 0 EUR
BIRBORB43726-100
Anti-CD28 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR