Anti-CD5 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB43741-100EA 0 EUR
BIRBORB43741-100
Anti-CD5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR