Anti-FOLH1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB43796-0.1
BIRBORB43796-0.1EA 0 EUR
BIRBORB43796-0.1
Anti-FOLH1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR