Anti-CD40 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB43825-0.1EA 0 EUR
BIRBORB43825-0.1
Anti-CD40 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR