Anti-ITGB1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44007-0.1EA 0 EUR
BIRBORB44007-0.1
Anti-ITGB1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR