Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB44080-100
BIRBORB44080-100EA 0 EUR
BIRBORB44080-100
Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR