Anti-CD177 Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB44117-100
BIRBORB44117-100EA 0 EUR
BIRBORB44117-100
Anti-CD177 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: CD177 (CD177 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR