Anti-ICAM3 Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
ORB44127-0.1
BIRBORB44127-0.1EA 0 EUR
BIRBORB44127-0.1
Anti-ICAM3 Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ICAM3 (intercellular adhesion molecule 3)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR