Anti-PTPRC Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44271-100EA 0 EUR
BIRBORB44271-100
Anti-PTPRC Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR