Anti-CD45RA Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44333-100EA 0 EUR
BIRBORB44333-100
Anti-CD45RA Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR