Anti-TF Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB44475-0.1
BIRBORB44475-0.1EA 0 EUR
BIRBORB44475-0.1
Anti-TF Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TF (transferrin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR