Anti-Kappa (L chain) Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB44506-0.1
BIRBORB44506-0.1EA 0 EUR
BIRBORB44506-0.1
Anti-Kappa (L chain) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Kappa (L chain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR