Anti-FCGR1A Mouse monoclonal antibody

Supplier: Biorbyt
ORB44624-0.1
BIRBORB44624-0.1EA 0 EUR
BIRBORB44624-0.1
Anti-FCGR1A Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR