Anti-BCL2 Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB44689-100EA 0 EUR
BIRBORB44689-100
Anti-BCL2 Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR