Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB44706-100
BIRBORB44706-100EA 0 EUR
BIRBORB44706-100
Anti-SELL Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR