Anti-Kinesin Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB44767-0.1
BIRBORB44767-0.1EA 0 EUR
BIRBORB44767-0.1
Anti-Kinesin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Kinesin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR