Anti-CD44 Rat Monoclonal Antibody

Supplier: Biorbyt
ORB44819-0.1
BIRBORB44819-0.1EA 0 EUR
BIRBORB44819-0.1
Anti-CD44 Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR