Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
BIRBORB44839-0.1EA 0 EUR
BIRBORB44839-0.1
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR