Anti-Ly6G Rat Monoclonal Antibody

Supplier: Biorbyt
BIRBORB44841-0.1EA 0 EUR
BIRBORB44841-0.1
Anti-Ly6G Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Ly6G
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR