Anti-IgM Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Biorbyt
BIRBORB44862-0.1EA 0 EUR
BIRBORB44862-0.1
Anti-IgM Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgM (Human) (human IgM)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR