Anti-IgM Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB44863-0.1
BIRBORB44863-0.1EA 0 EUR
BIRBORB44863-0.1
Anti-IgM Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgM (Human) (human IgM)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR