Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
ORB44868-0.1
BIRBORB44868-0.1EA 0 EUR
BIRBORB44868-0.1
Anti-IgG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG (Human) (human IgG)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR