Anti-CDK13 Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB45256-100EA 0 EUR
BIRBORB45256-100
Anti-CDK13 Mouse monoclonal antibody
Antibodies
Western Blot: 1:500

Type: Primary
Antigen: CDK13 (cyclin-dependent kinase 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR