Anti-FOXO3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB47943-100EA 0 EUR
BIRBORB47943-100
Anti-FOXO3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOXO3 (forkhead box O3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR