Anti-MCL1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB47970-100EA 0 EUR
BIRBORB47970-100
Anti-MCL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MCL1 (myeloid cell leukemia 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR