Anti-XIAP Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB47979-100
BIRBORB47979-100EA 0 EUR
BIRBORB47979-100
Anti-XIAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR