Anti-CALM1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB47986-100EA 0 EUR
BIRBORB47986-100
Anti-CALM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CALM1 (calmodulin 1 (phosphorylase kinase, delta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR