Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48000-100EA 0 EUR
BIRBORB48000-100
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1B (interleukin 1, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR