Anti-BCAT1 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48005-100
BIRBORB48005-100EA 0 EUR
BIRBORB48005-100
Anti-BCAT1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: BCAT1 (branched chain amino-acid transaminase 1, cytosolic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR