Anti-KITLG Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48031-100
BIRBORB48031-100EA 0 EUR
BIRBORB48031-100
Anti-KITLG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KITLG (KIT ligand)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR