Anti-c-Myc tag Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB48094-100 ORB48094-20
BIRBORB48094-100EA 0 EUR
BIRBORB48094-100 BIRBORB48094-20
Anti-c-Myc tag Mouse Monoclonal Antibody
Antibodies
  • Western Blot: 1:3000-1:10000; Immunofluorescence: 1:200

Type: Primary
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR