Anti-AKR1B1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48276-100EA 0 EUR
BIRBORB48276-100
Anti-AKR1B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AKR1B1 (aldo-keto reductase family 1, member B1 (aldose reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR