Anti-BRCA1 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48292-100
BIRBORB48292-100EA 0 EUR
BIRBORB48292-100
Anti-BRCA1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR