Anti-CA2 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48298-100EA 0 EUR
BIRBORB48298-100
Anti-CA2 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR