Anti-CD79B Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48314-100
BIRBORB48314-100EA 0 EUR
BIRBORB48314-100
Anti-CD79B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR