Anti-CDC25C Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48318-100
BIRBORB48318-100EA 0 EUR
BIRBORB48318-100
Anti-CDC25C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR