Anti-CDKN3 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48325-100EA 0 EUR
BIRBORB48325-100
Anti-CDKN3 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CDKN3 (cyclin-dependent kinase inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR