Anti-DNMT3A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48354-100EA 0 EUR
BIRBORB48354-100
Anti-DNMT3A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR